2-Ethoxy-4-fluor-1-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1019-5G AN-1019-1G AN-1019-25G
COBBAN-1019-5GEA 352 EUR
COBBAN-1019-5G COBBAN-1019-1G COBBAN-1019-25G
2-Ethoxy-4-fluor-1-nitrobenzol
2-Ethoxy-4-fluor-1-nitrobenzol
Formel: C₈H₈FNO₃
Molecular Weight: 185,1524 g/mol
MDL: MFCD04115630
CAS-Nummer: 28987-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR