4-Fluor-2-nitro-1-(trifluormethoxy)benzol

Lieferant: COMBI-BLOCKS

AN-1446-1G AN-1446-5G
COBBAN-1446-1GEA 0 EUR
COBBAN-1446-1G COBBAN-1446-5G
4-Fluor-2-nitro-1-(trifluormethoxy)benzol
4-Fluor-2-nitro-1-(trifluormethoxy)benzol
Formel: C₇H₃F₄NO₃
MDL: MFCD12025954
CAS-Nummer: 123572-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR