(R)-(+)-N,N-Dimethyl-1-phenylethanamin

Lieferant: COMBI-BLOCKS

Synonyme: (R)-N,N-Dimethyl-1-phenylethanamine

OS-7510-5G OS-7510-25G OS-7510-1G
COBBOS-7510-5GEA 204 EUR
COBBOS-7510-5G COBBOS-7510-25G COBBOS-7510-1G
(R)-(+)-N,N-Dimethyl-1-phenylethanamin
(R)-(+)-N,N-Dimethyl-1-phenylethanamin
Formel: C₁₀H₁₅N
Molecular Weight: 149,230 g/mol
MDL: MFCD00008857
CAS-Nummer: 19342-01-9
EINECS: 242-976-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR