L(+)-Isoleucinol

Lieferant: COMBI-BLOCKS

Synonyme: L-Isoleucinol, (S)-(+)-Isoleucinol, (2S,3S)-2-Amino-3-methyl-1-pentanol, (2S,3S)-2-Amino-3-methylpentan-1-ol

HC-6498-25G HC-6498-5G HC-6498-1G
COBBHC-6498-25GEA 130 EUR
COBBHC-6498-25G COBBHC-6498-5G COBBHC-6498-1G
L(+)-Isoleucinol
L(+)-Isoleucinol
Formel: C₆H₁₅NO
Molecular Weight: 117.19 g/mol
MDL: MFCD00004731
CAS-Nummer: 24629-25-2
EINECS: 246-371-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR