2-Ethoxy-4-methylphenol 95%
Lieferant: Thermo Scientific
|
Achtung
|
L08898.09
L08898.04
L08898.09EA
275
EUR
L08898.09
L08898.04
2-Ethoxy-4-methylphenol 95%
2-Ethoxy-4-methylphenol
Purity: 95%
CAS No.: 2563-07-7
Molecular Formula: C9H12O2
Formula Weight: 152.19
Refractive Index: 1.5230
Melting Point: 28-30°
Boiling Point: 103°/3mm
Beilstein Reference: 1865448
MDL No.: MFCD00043697
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Siedepunkt: 103 °C (3 mmHg) Schmelzpunkt: 28…30 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043697 CAS-Nummer: 2563-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...