1-Isopropylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Isopropyl-1H-imidazole

OR-2366-25G OR-2366-5G
COBBOR-2366-25GEA 34.4 EUR
COBBOR-2366-25G COBBOR-2366-5G
1-Isopropylimidazol
1-Isopropylimidazol
Formel: C₆H₁₀N₂
Molecular Weight: 110,1600 g/mol
MDL: MFCD00234239
CAS-Nummer: 4532-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR