1-Tritylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
Clotrimazole Impurity F, 1-Trityl-1H-imidazole
HI-1042-25G
HI-1042-1G
HI-1042-100G
HI-1042-5G
COBBHI-1042-25GEA
264
EUR
COBBHI-1042-25G
COBBHI-1042-1G
COBBHI-1042-100G
COBBHI-1042-5G
1-Tritylimidazol
1-Tritylimidazol
Formel:
C₂₂H₁₈N₂ Molecular Weight: 310,40 g/mol |
MDL:
MFCD00229427 CAS-Nummer: 15469-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...