1,5-Dimethyl-1H-pyrazol
Lieferant: COMBI-BLOCKS
HI-1486-5G
HI-1486-1G
COBBHI-1486-5GEA
58.5
EUR
COBBHI-1486-5G
COBBHI-1486-1G
1,5-Dimethyl-1H-pyrazol
1,5-Dimethyl-1H-pyrazole
Formel:
C₅H₈N₂ Molecular Weight: 96,1300 g/mol |
MDL:
MFCD01646129 CAS-Nummer: 694-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...