N-Acetyl-5-brom-2-fluoranilin
Lieferant: COMBI-BLOCKS
Synonyme:
5'-Bromo-2'-fluoroacetanilide
HC-2836-5G
HC-2836-1G
COBBHC-2836-5GEA
0
EUR
COBBHC-2836-5G
COBBHC-2836-1G
N-Acetyl-5-brom-2-fluoranilin
N-Acetyl-5-brom-2-fluoranilin
Formel:
C₈H₇BrFNO |
MDL:
MFCD12546480 CAS-Nummer: 88288-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...