2-Methylbutyrylchlorid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
DL-2-Methylbutyryl chloride
B22599.06
B22599.14
B22599.22
B22599.06EA
43.4
EUR
B22599.06
B22599.14
B22599.22
2-Methylbutyrylchlorid ≥99%
2-Methylbutyrylchlorid
Purity: 99%
CAS No.: 57526-28-0
Molecular Formula: C5H9ClO
Formula Weight: 120.58
Flash Point: 20°(68°F)
Refractive Index: 1.4180
Boiling Point: 116-117°
UN No.: 2924
MDL No.: MFCD00041726
Formel:
C₅H₉ClO Molecular Weight: 120.58 g/mol Siedepunkt: 116…117 °C Dichte: 0.98 Flammpunkt: 20 °C (68 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041726 CAS-Nummer: 57526-28-0 EINECS: 260-787-3 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...