tert-Butylacetylchlorid 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,3-Dimethylbutyryl chloride

107790500 107792500 107790100
ACRO107790500EA 80.6 EUR
ACRO107790500 ACRO107792500 ACRO107790100
tert-Butylacetylchlorid 99%
tert-Butylacetylchlorid
Formel: C₆H₁₁ClO
Molecular Weight: 134,61 g/mol
Siedepunkt: 127…129 °C (760 mmHg)
Dichte: 0,96 g/cm³
Flammpunkt: 21 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00000737
CAS-Nummer: 7065-46-5
EINECS: 230-356-4
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4230 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
Titration RCOOH ≤1 %
pH 3.8 to 7.6 (Ph. Eur.(c=1, about 55°C))
Arsenic (As) ≤1 ppm
Lead (Pb) ≤5 ppm
Heavy metals (as Pb) ≤50 ppm
Sulfur dioxide (SO2) ≤40 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR