3'-Brom-5'-fluoracetophenon 96%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 1-(3-Bromo-5-fluorophenyl)ethanone
H62977.03 H62977.06
H62977.03EA 128 EUR
H62977.03 H62977.06
3'-Brom-5'-fluoracetophenon 96%
3'-Brom-5'-fluoracetophenon
Purity: 96% CAS No.: 105515-20-6 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847746
CAS-Nummer: 105515-20-6