4-(Dichlormethyl)benzonitril

Lieferant: CHEMPUR

CD08949.250 CD08949.1
CHMPCD08949.250EA 0 EUR
CHMPCD08949.250 MAYBCD08949.1
4-(Dichlormethyl)benzonitril
4-(Dichlormethyl)benzonitril
Formel: C₈H₅Cl₂N
Molecular Weight: 186,0400 g/mol
MDL: MFCD00045606
CAS-Nummer: 74231-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR