2,4-Dichlor-5-fluorbenzylalkohol

Lieferant: FLUOROCHEM

005730-1G 005730-5G
FLUO005730-1GEA 63.9 EUR
FLUO005730-1G FLUO005730-5G
2,4-Dichlor-5-fluorbenzylalkohol
2,4-Dichlor-5-fluorbenzylalkohol
Formel: C₇H₅Cl₂FO
MDL: MFCD01631380
CAS-Nummer: 219765-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR