3-Brom-6-chlorpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-6-chloropyridine-2-carboxylic acid

PY-7024-5G PY-7024-1G
COBBPY-7024-5GEA 71.7 EUR
COBBPY-7024-5G COBBPY-7024-1G
3-Brom-6-chlorpicolinsäure
3-Brom-6-chlorpicolinsäure
Formel: C₆H₃BrClNO₂
Molecular Weight: 236.4505 g/mol
MDL: MFCD09258777
CAS-Nummer: 929000-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR