6-Amino-3-brompicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-Amino-3-bromo-2-pyridinecarboxylic acid

CA-4181-5G CA-4181-25G CA-4181-1G
COBBCA-4181-5GEA 645 EUR
COBBCA-4181-5G COBBCA-4181-25G COBBCA-4181-1G
6-Amino-3-brompicolinsäure
6-Amino-3-brompicolinsäure
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,0200 g/mol
MDL: MFCD10699642
CAS-Nummer: 1033201-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR