5-Hydroxypicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-hydroxypyridine-2-carboxylic acid

CA-4311-5G CA-4311-1G
COBBCA-4311-5GEA 63.2 EUR
COBBCA-4311-5G COBBCA-4311-1G
5-Hydroxypicolinsäure
5-Hydroxypicolinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
MDL: MFCD00661309
CAS-Nummer: 15069-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR