Cyclobutylphenylketon

Lieferant: CHEMPUR

Synonyme: Cyclobutyl Phenyl Ketone

FL3371.5 FL3371.25
CHMPFL3371.5EA 0 EUR
CHMPFL3371.5 CHMPFL3371.25
Cyclobutylphenylketon
Cyclobutylphenylketon
Formel: C₁₁H₁₂O
Molecular Weight: 160,2124 g/mol
MDL: MFCD00001326
CAS-Nummer: 5407-98-7
EINECS: 226-473-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR