1-(3-Methoxypropyl)piperazin

Lieferant: FLUOROCHEM

009805-1G 009805-5G
FLUO009805-1GEA 72.9 EUR
FLUO009805-1G FLUO009805-5G
1-(3-Methoxypropyl)piperazin
1-(3-Methoxypropyl)piperazin
Formel: C₈H₁₈N₂O
MDL: MFCD02093537
CAS-Nummer: 88708-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR