Blei-Zirkonium-Titan-Oxid (∼30 wt% Oxide) precursor_polymeric
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Lead zirconate titanate
39758.09 39758.18
39758.09EA 107 EUR
39758.09 39758.18
Blei-Zirkonium-Titan-Oxid (∼30 wt% Oxide) precursor_polymeric
Lead zirconium titanium oxide
CAS No.: 12626-81-2 Physical Form: Semi-solid/viscous liquid Appearance: Pale yellow Solubility: Soluble in alcohols UN No.: 1993 MDL No.: MFCD00212270
Used in piezoelectric devices and electro-optic devices
Formel: PbZr(1-x)TixO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00212270
CAS-Nummer: 12626-81-2
EINECS: 235-727-4
UN: 1993
ADR: 3 III