1-(2,5-Dibromphenyl)thioharnstoff
Lieferant: COMBI-BLOCKS
HC-3368-5G
HC-3368-1G
COBBHC-3368-5GEA
303
EUR
COBBHC-3368-5G
COBBHC-3368-1G
1-(2,5-Dibromphenyl)thioharnstoff
1-(2,5-Dibromphenyl)thioharnstoff
Formel:
C₇H₆Br₂N₂S |
MDL:
MFCD14673413 CAS-Nummer: 854890-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...