2-Brom-6-nitrobenzylalkohol

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-6-nitrophenyl)methanol

SS-3693-5G SS-3693-1G
COBBSS-3693-5GEA 412 EUR
COBBSS-3693-5G COBBSS-3693-1G
2-Brom-6-nitrobenzylalkohol
2-Brom-6-nitrobenzylalkohol
Formel: C₇H₆BrNO₃
Molecular Weight: 232,0300 g/mol
MDL: MFCD11036298
CAS-Nummer: 861106-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR