2-(2,4-Dichlorphenoxy)propionsäurehydrazid ≥98%

Lieferant: FLUOROCHEM

011143-25G 011143-100G
FLUO011143-25GEA 141 EUR
FLUO011143-25G FLUO011143-100G
2-(2,4-Dichlorphenoxy)propionsäurehydrazid ≥98%
2-(2,4-Dichlorphenoxy)propionsäurehydrazid
Formel: C₉H₁₀Cl₂N₂O₂
Molecular Weight: 249,09 g/mol
MDL: MFCD00204152
CAS-Nummer: 15253-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR