2-(2,4-Dichlorphenoxy)propionsäurehydrazid ≥98%
Lieferant: MENAI
ST2300.25
ST2300.100
MENAST2300.25EA
0
EUR
MENAST2300.25
MENAST2300.100
2-(2,4-Dichlorphenoxy)propionsäurehydrazid ≥98%
2-(2,4-Dichlorphenoxy)propionsäurehydrazid
Formel:
C₉H₁₀Cl₂N₂O₂ Molecular Weight: 249,09 g/mol |
MDL:
MFCD00204152 CAS-Nummer: 15253-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...