3-(3-Chlorphenoxy)propionsäure

Lieferant: CHEMPUR

05-0039.1 05-0039.5
CHMP05-0039.1EA 0 EUR
CHMP05-0039.1 CHMP05-0039.5
3-(3-Chlorphenoxy)propionsäure
3-(3-Chlorphenoxy)propionsäure
Formel: C₉H₉ClO₃
MDL: MFCD00151673
CAS-Nummer: 7170-50-5
EINECS: 230-522-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR