5-Amino-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-2-pyridinecarbonitrile, 5-Amino-2-cyanopyridine

M-Clarity™ Qualitätsstufe = MQ100

538906-100G 538906-25G
SIAL538906-100GEA 482 EUR
SIAL538906-100G SIAL538906-25G
5-Amino-2-pyridincarbonitril, Sigma-Aldrich®
5-Amino-2-pyridincarbonitril
Formel: C₆H₅N₃
Molecular Weight: 119,1240 g/mol
MDL: MFCD03093956
CAS-Nummer: 55338-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR