1-(2-(Trifluormethyl)phenyl)thioharnstoff

Lieferant: MENAI

Synonyme: N-[2-(Trifluoromethyl)phenyl]thiourea

SR469.5 SR469.10
MENASR469.5EA 0 EUR
MENASR469.5 MENASR469.10
1-(2-(Trifluormethyl)phenyl)thioharnstoff
1-(2-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220,22 g/mol
MDL: MFCD00041161
CAS-Nummer: 1736-71-6
EINECS: 217-084-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR