1-(3-(Trifluormethyl)phenyl)thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N-[3-(Trifluoromethyl)phenyl]thiourea

009390-25G 009390-5G
FLUO009390-25GEA 155 EUR
FLUO009390-25G FLUO009390-5G
1-(3-(Trifluormethyl)phenyl)thioharnstoff
1-(3-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220,22 g/mol
MDL: MFCD00041171
CAS-Nummer: 1736-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR