1-(3-(Trifluormethyl)phenyl)thioharnstoff

Lieferant: MENAI

Synonyme: N-[3-(Trifluoromethyl)phenyl]thiourea

ER19.5 ER19.25 ER19.100 ER19.200
MENAER19.5EA 0 EUR
MENAER19.5 MENAER19.25 MENAER19.100 MENAER19.200
1-(3-(Trifluormethyl)phenyl)thioharnstoff
1-(3-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220.22 g/mol
MDL: MFCD00041171
CAS-Nummer: 1736-70-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR