1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
Lieferant: Thermo Scientific
Synonyme:
N-[3,5-Bis(trifluoromethyl)phenyl]thiourea
SPB03351.1
SPB03351.10
SPB03351.50
MAYBSPB03351.1EA
0
EUR
MAYBSPB03351.1
MAYBSPB03351.10
MAYBSPB03351.50
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
Formel:
C₉H₆F₆N₂S Molecular Weight: 288,22 g/mol |
MDL:
MFCD00041172 CAS-Nummer: 175277-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...