1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff

Lieferant: MENAI

Synonyme: N-[3,5-Bis(trifluoromethyl)phenyl]thiourea

LH258.5 LH258.25 LH258.100
MENALH258.5EA 0 EUR
MENALH258.5 MENALH258.25 MENALH258.100
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
Formel: C₉H₆F₆N₂S
Molecular Weight: 288,22 g/mol
MDL: MFCD00041172
CAS-Nummer: 175277-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR