1-(4-(Trifluormethyl)phenyl)thioharnstoff, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-[4-(Trifluoromethyl)phenyl]thiourea

M-Clarity™ Qualitätsstufe = MQ100

654760-1G 654760-5G
SIAL654760-1GEA 38.6 EUR
SIAL654760-1G SIAL654760-5G
1-(4-(Trifluormethyl)phenyl)thioharnstoff, Sigma-Aldrich®
1-(4-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220,22 g/mol
MDL: MFCD00041189
CAS-Nummer: 1736-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR