3,5-Difluor-N-methylanilin

Lieferant: COMBI-BLOCKS

AN-1248-1G AN-1248-5G
COBBAN-1248-1GEA 25.5 EUR
COBBAN-1248-1G COBBAN-1248-5G
3,5-Difluor-N-methylanilin
3,5-Difluor-N-methylanilin
Formel: C₇H₇F₂N
MDL: MFCD09878315
CAS-Nummer: 470458-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR