4-Iod-2-pyridincarbonitril

Lieferant: COMBI-BLOCKS

CA-4253-5G CA-4253-1G
COBBCA-4253-5GEA 1700 EUR
COBBCA-4253-5G COBBCA-4253-1G
4-Iod-2-pyridincarbonitril
4-Iod-2-pyridincarbonitril
Formel: C₆H₃IN₂
MDL: MFCD08689497
CAS-Nummer: 913836-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR