m-Xylylendiamin 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,3-Benzenedimethanamine
141020050
141021000
141025000
ACRO141020050EA
26.3
EUR
ACRO141020050
ACRO141021000
ACRO141025000
m-Xylylendiamin 99%
m-Xylylendiamin
Formel:
C₈H₁₂N₂ Molecular Weight: 136,1943 g/mol Siedepunkt: 265 °C (745 mmHg) Dichte: 1,03 g/cm³ Flammpunkt: >112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008119 CAS-Nummer: 1477-55-0 EINECS: 216-032-5 UN: 2735 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5700 to 1.5720 (20°C, 589 nm) |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...