m-Xylylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,3-Benzenedimethanamine
M-Clarity™ Qualitätsstufe = MQ100
X1202-100G
X1202-500G
X1202-5G
SIALX1202-100GEA
34.5
EUR
SIALX1202-100G
SIALX1202-500G
SIALX1202-5G
m-Xylylendiamin, Sigma-Aldrich®
m-Xylylendiamin
Formel:
C₈H₁₂N₂ Molecular Weight: 136,1943 g/mol |
MDL:
MFCD00008119 CAS-Nummer: 1477-55-0 EINECS: 216-032-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...