p-Xylylendiamin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1,4-Bis(aminomethyl)benzene, 1,4-Phenylenedimethanamine

L02995.04 L02995.09 L02995.18 AB177666.0050
L02995.04EA 43.7 EUR
L02995.04 L02995.09 L02995.18 ABCRAB177666.0050
p-Xylylendiamin 97%
p-Xylylendiamin

Purity: 97% CAS No.: 539-48-0 Molecular Formula: C8H12N2 Formula Weight: 136.20 Flash Point: 134°(273°F) Melting Point: 59-63° Boiling Point: 230°/10mm Beilstein Reference: 2206190 UN No.: 3259 MDL No.: MFCD00009821

Formel: C₈H₁₂N₂
Molecular Weight: 136,20 g/mol
Siedepunkt: 230 °C (10 mmHg)
Schmelzpunkt: 59…63 °C
Flammpunkt: 134 °C (273 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009821
CAS-Nummer: 539-48-0
EINECS: 208-719-3
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR