p-Xylylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,4-Bis(aminomethyl)benzene, 1,4-Phenylenedimethanamine
M-Clarity™ Qualitätsstufe = MQ200
279633-25G
279633-5G
SIAL279633-25GEA
387
EUR
SIAL279633-25G
SIAL279633-5G
p-Xylylendiamin, Sigma-Aldrich®
p-Xylylendiamin
Formel:
C₈H₁₂N₂ Molecular Weight: 136,20 g/mol |
MDL:
MFCD00009821 CAS-Nummer: 539-48-0 EINECS: 208-719-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...