4-(Diethylamino)azobenzol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N,N-Diethyl-4-(phenyldiazenyl)aniline

B23915.06 B23915.14 B23915.22
B23915.06EA 30.4 EUR
B23915.06 B23915.14 B23915.22
4-(Diethylamino)azobenzol 98%
4-(Diethylamino)azobenzol

Purity: 98% CAS No.: 2481-94-9 Molecular Formula: C16H19N3 Formula Weight: 253.35 Melting Point: 93-96° MDL No.: MFCD00009043

Formel: C₁₆H₁₉N₃
Molecular Weight: 253,35 g/mol
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009043
CAS-Nummer: 2481-94-9
EINECS: 219-616-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR