Diethylmethylvinylsilan

Lieferant: CHEMPUR SILANE

SILS06450.10 SILS06450.25
SILNSILS06450.10EA 0 EUR
SILNSILS06450.10 SILNSILS06450.25
Diethylmethylvinylsilan
Diethylmethylvinylsilan
Formel: C₇H₁₆Si
MDL: MFCD00041344
CAS-Nummer: 18292-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR