5-Chlor-2-fluor-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Chloro-2-fluoro-4-methylphenyl)boronic acid

BB-8514-5G BB-8514-1G
COBBBB-8514-5GEA 1490 EUR
COBBBB-8514-5G COBBBB-8514-1G
5-Chlor-2-fluor-4-methylphenylboronsäure
5-Chlor-2-fluor-4-methylphenylboronsäure
Formel: C₇H₇BClFO₂
Molecular Weight: 188,3900 g/mol
MDL: MFCD07368241
CAS-Nummer: 1072952-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR