5-Butylpicolinsäure 99%

Lieferant: Thermo Fisher Scientific
Achtung

198960010 198962500
ACRO198960010EA 237 EUR
ACRO198960010 ACRO198962500
5-Butylpicolinsäure 99%
5-Butylpicolinsäure
Formel: C₁₀H₁₃NO₂
Schmelzpunkt: 97…101 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006298
CAS-Nummer: 536-69-6
EINECS: 208-643-0
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 101 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR