Eriochromblauschwarz B
Lieferant: Thermo Scientific
|
Achtung
|
J63396.22
J63396.36
J63396.22EA
28
EUR
J63396.22
J63396.36
Eriochromblauschwarz B
Eriochromblauschwarz B
CAS No.: 3564-14-5
Molecular Formula: C20H13N2NaO5S
Formula Weight: 416.39
Physical Form: Powder
Appearance: Brown to black
Formel:
C₂₀H₁₃N₂NaO₅S Molecular Weight: 416,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003934 CAS-Nummer: 3564-14-5 EINECS: 222-639-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...