2-Acetylbenzonitril

Lieferant: BIONET RESEARCH

Synonyme: 2-Acetylbenzenecarbonitrile

1T-0269.500 1T-0269.1
BIOT1T-0269.500EA 0 EUR
BIOT1T-0269.500 BIOT1T-0269.1
2-Acetylbenzonitril
2-Acetylbenzonitril
Formel: C₉H₇NO
Molecular Weight: 145,1600 g/mol
MDL: MFCD00272571
CAS-Nummer: 91054-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR