(1R,2R)-trans-2-Aminocyclohexanol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(1R,2R)-2-Aminocyclohexanol hydrochloride
M-Clarity™ Qualitätsstufe = MQ100
671762-1G
671762-250MG
SIAL671762-1GEA
178
EUR
SIAL671762-1G
SIAL671762-250MG
(1R,2R)-trans-2-Aminocyclohexanol Hydrochlorid, Sigma-Aldrich®
(1R,2R)-trans-2-Aminocyclohexanol Hydrochlorid
Formel:
C₆H₁₄ClNO Molecular Weight: 151,6345 g/mol |
MDL:
MFCD09259962 CAS-Nummer: 13374-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...