4-Phenyl-3-butin-2-ol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

669288-25G 669288-5G
SIAL669288-25GEA 263 EUR
SIAL669288-25G SIAL669288-5G
4-Phenyl-3-butin-2-ol, Sigma-Aldrich®
4-Phenyl-3-butin-2-ol
Formel: C₁₀H₁₀
Molecular Weight: 146.19 g/mol
MDL: MFCD00046651
CAS-Nummer: 5876-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR