1-Methyl-5-nitroimidazol, Sigma-Aldrich®

Lieferant: Merck

94489-1G-F 94489-5G-F
SIAL94489-1G-FEA 217 EUR
SIAL94489-1G-F SIAL94489-5G-F
1-Methyl-5-nitroimidazol, Sigma-Aldrich®
1-Methyl-5-nitroimidazole
Formel: C₄H₅N₃O₂
Molecular Weight: 127,1000 g/mol
MDL: MFCD00126968
CAS-Nummer: 3034-42-2
EINECS: 221-225-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR