4-Brom-2-chloranisol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2-chloro-1-methoxybenzene

OT-0527-1G OT-0527-5G
COBBOT-0527-1GEA 25.5 EUR
COBBOT-0527-1G COBBOT-0527-5G
4-Brom-2-chloranisol
4-Brom-2-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,4789 g/mol
MDL: MFCD00128076
CAS-Nummer: 50638-47-6
EINECS: 256-672-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR