1-Brom-3-(trifluormethylthio)benzol

Lieferant: CHEMPUR

FL8013.5 FL8013.10 008013-1G 008013-5G
CHMPFL8013.5EA 0 EUR
CHMPFL8013.5 CHMPFL8013.10 FLUO008013-1G FLUO008013-5G
1-Brom-3-(trifluormethylthio)benzol
1-Bromo-3-(trifluoromethylthio)benzene
Formel: C₇H₄BrF₃S
MDL: MFCD00236348
CAS-Nummer: 2252-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR