1,2,4-Trifluor-5-(methylsulfonyl)benzol

Lieferant: COMBI-BLOCKS

OT-0831-1G OT-0831-250M
COBBOT-0831-1GEA 0 EUR
COBBOT-0831-1G COBBOT-0831-250M
1,2,4-Trifluor-5-(methylsulfonyl)benzol
1,2,4-Trifluor-5-(methylsulfonyl)benzol
Formel: C₇H₅F₃O₂S
MDL: MFCD12026059
CAS-Nummer: 845616-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR